Hose Fittings, Band Clamps, for 2" OD Hose, 25/Pk
Supplier: VWR International
Quick and easy assembly with band clamp tool.
- Constructed of durable stainless steel
Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.