Masterflex® Liquid Flow Switch for High Inline Pressures, Brass; 0.5 GPM
Supplier: VWR International
VWR Item#MFLX32774-32
Your Organization's Price:
Retrieving
Ideal for high pressure applications.
  • Withstands up to 1500 psi
  • Effortlessly install with 3/8" NPT(M) connections
The one-piece magnetic PPS composite piston makes these normally open switches ideal for high pressure applications. The single pole single throw (SPST) switches withstand up to 1500 psi at 70 °F (21 °C)! Use a 100 µm filter to protect your switch. Switches include 24 to 26" lead wires.