Hedland H205B-020 Water Flowmeter, 0.2-2.0 GPM, Brass, 1/4" NPT(F)
Supplier: VWR International
VWR Item#MFLX32509-56
Your Organization's Price:
Retrieving
Easy to read scale.
  • Direct reading
  • Install in any position
  • 360º Rotatable Guard/Scale
  • Easier ot Read Linear Scale
  • No Flow Straighteners or Special Piping Required
  • Insensitive to Shock and Vibration
  • Good Viscosity Stability
  • Temperature up to 240ºF
  • Accuracy ±2% Full Scale
  • Repeatability ±1%
  • Special Scales Available
Ideal for applications that are sensitive to shock and vibration.