Masterflex® Fitting, 316 Stainless Steel, Straight, Nipple Thread Union, 3/4" NPT(M)
Supplier: VWR International
VWR Item#MFLX31410-44
Your Organization's Price:
Retrieving
Deigned for permanent connection.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/4" NPT(M)
Feature rolled threads for improved strength. Very good chemical resistance. Rated to 7500 psi at 77 °F ( 25 °C). Usable from -65 to 550 °F (-53 to 289 °C).