Masterflex® Transfer Tubing, C-Flex® ADCF, 3/8" ID x 5/8" OD; 350 Ft
Supplier: VWR International
Meets the demands of various industries.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimize downtime
- Tubing size: ³/₈" int.Ø × ¹/₂" out.Ø
<B>Characteristics:</B> Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Transparent.<br /><B>Certification:</B> Resin meets USP and Class VI requirements. Tubing is manufactured according to GMP.<br /><B>Temperature range:</B> -50 to 275 °F (-45 to 135 °C)<br /><B>Sterilize:</B> Autoclave, ethylene oxide, or gamma irradiation.