PVC Stopcock with Flow Indication, 1/4" NPT(F)×1/4" NPT(F)
Supplier: VWR International
Easily control flow with flow indicator handle.
- Constructed of durable PVC and EPDM
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120 °F (49 °C); maximum vacuum is 29.9" Hg.