Masterflex® Brass Metering Valve, 90 deg flow, 350 mL/min
Supplier: VWR International
VWR Item#MFLX03218-78
Your Organization's Price:
Retrieving
Choose from different metal materials based on your application.
  • Brass and 316 Stainless Steel material options available
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.