Masterflex® Fittings, HDPE, Elbow, Hose Barb Union, 3/32" ID; 100/PK
Supplier: VWR International
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/16" ID
Create 90° bends in flexible tubing—without the kinks!