Masterflex® Fitting, HDPE, Tee, Hosebarb to Thread Adapter, 1/4" ID x 1/4" ID X 1/4" NPT(M); 10/PK
Supplier: VWR International
VWR Item#MFLX30624-04
Your Organization's Price:
Retrieving
Quick and easy assembly.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/4" ID x 1/4" ID X 1/4" NPT(M)
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.