Masterflex® Ismatec® Pump Tubing, Tygon® S3™ E-LFL, 2.62 mm ID; 100 ft
Supplier: VWR International
VWR Item#MFLX06449-47
Your Organization's Price:
Retrieving
Longest life of all Tygon® formulations.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Tubing Inner Diameter: 2.62 mm
<B>Applications</B>: General laboratory use<br /><B>Characteristics</B>: Long life, won’t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, transparent