Masterflex® Fitting, PVDF, Straight, Hose Barb Plug , 1/16" ID; 10/PK
Supplier: VWR International
VWR Item#MFLX40615-38
Your Organization's Price:
Retrieving
Protect against contaminants in tubing lines.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/16" ID
Use these barbed press-in-plugs to keep open tubing lines or tubing assemblies free of foreign contaminants and ensure process purity. Plugs are manufactured from 100% virgin materials in a ISO Class 8 (formerly Class 100,000) environment and include full material traceability. Available in PVDF or nylon. All plugs are sterilizable by gamma irradiation; nylon plugs may also be autoclaved.