Masterflex® Fitting, PVDF, Straight, Hose Barb Reducer, 5/8" ID x 5/16" ID; 10/PK
Supplier: VWR International
VWR Item#MFLX40614-70
Your Organization's Price:
Retrieving
Best for low-pressure applications.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range -80 to 265 °F (-62 to 129 °C)
  • Connection Type: 1/2" ID x 3/8" ID