Masterflex® L/S® Spooled High-Performance Precision Pump Tubing, BioPharm Platinum-Cured Silicone, L/S 24; 400 ft
Supplier: VWR International
VWR Item#MFLX96423-24
Your Organization's Price:
Retrieving
Ultra-smooth inner surface minimizes particle entrapment.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 24
<B>Applications</B>: Pharmaceutical, cosmetic, research, and auto-analysis applications<br /><B>Characteristics</B>: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent<br />Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.