Masterflex® L/S® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, L/S 36; 25 ft
Supplier: VWR International
VWR Item#MFLX96410-36
Your Organization's Price:
Retrieving
Excellent biocompatibility that meets multiple classifications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Unique packaging options include packs, cases and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
  • Tubing Size: L/S 36
<B>Applications</B>: Pharmaceutical, biotech, peristaltic pumping, and food applications<br /><B>Characteristics</B>: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
See 96119-15 for the irradiated version of this tubing.