Masterflex® B/T® PerfectPosition® Pump Tubing, Peroxide-Cured Silicone, B/T 91; 10 ft
Supplier: VWR International
VWR Item#MFLX96400-91
Your Organization's Price:
Retrieving
Ensure optimum performance from your Masterflex® pump.
  • Excellent biocompatibility
  • No additives, plasticizers or DOP; odorless and nontoxic, fungus-resistant
  • No taste imparted to transported fluids
  • Extremely good at low temperatures
  • Weather, ozone, corona, and radiation resistant
  • Minimal tendency to take a set
  • PerfectPosition™ tubing retention marks indicate the exact placement of tubing in the pump head to provide the best performance and life of the tubing
  • Custom extruded to fit 77110- and 77111-series Masterflex® B/T® pumps and pump heads
  • Engineered for long life in peristaltic pump applications
Characteristics: Superior heat resistance, low compression set, odorless, and nontoxic. Translucent. <br />Certifications: Elastomer is FDA-compliant (21 CFR 177.2600). Tubing is <br />manufactured according to GMP. <br />Temperature range: –60 to 460 °F (–51 to 238 °C) <br />Sterilize: Autoclave or gamma irradiation.