ChemFluor FEP Tubing, 1/16" ID×1/8" OD; 50 ft/Pk
Supplier: Saint Gobain Life Sciences
Meets the demands of various industries.
- Ideal for general-purpose, single-use, and extended fluid path applications
<B>Applications:</B> Transfer of abrasive fluids; pharmaceutical processes.<br /><B>Characteristics:</B> Good chemical resistance at a low cost, temperature resistant, low coefficient of friction, semi-flexible. Transparent.<br /><B>Certification:</B>FDA compliant.<br /><B>Temperature range:</B>–454 to 400 °F (–270 to 205 °C<br /><B>Sterilize:</B>With autoclave, ethylene oxide, or use dry heat.