Masterflex® Ismatec® Pump Tubing, Tygon® E-Lab, 0.38 mm ID; 100 ft
Supplier: VWR International
VWR Item#MFLX06460-14
Your Organization's Price:
Retrieving
Ideal for general transfer applications.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Tubing Inner Diameter: 0.38 mm
<B>Applications</B>: General laboratory use<br /><B>Characteristics</B>: Nontoxic, nonaging, nonoxidizing, easy flow monitoring, transparent<br />The overall length of E-Lab 06460-series extension tubing is 100 feet (30 m). The legacy Ismatec extension tubing was 10 m long.