Masterflex® I/P® High-Performance Precision Pump Tubing, Tygon® E-Food, I/P 88; 50 ft
Supplier: VWR International
VWR Item#MFLX06418-88
Your Organization's Price:
Retrieving
The preferred tubing of the food industry.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Smooth, nonporous inner bore inhibits particle entrapment and minimizes microbial growth
  • I/P® 88 tubing has a ¹/₂” (12.7 mm) inner diameter
  • Accepts ¹/₂” hose barb for tubing connection
Masterflex® Tygon® E-Food High-Performance Precision pump tubing is nonwetting, odorless, tasteless, and nontoxic making it ideal for the food and beverage industry. This tubing is resistant to harsh alkaline clears and commonly used sanitizers. Ideal for food and beverage, milk, and dairy applications, this tubing complies with FDA, EU Foods, NSF, and 3-A standards for contact with food products.