Masterflex® Transfer Tubing, Versilon™ A-60-G, ¾" ID × 1" OD, 50 ft.
Supplier: VWR International
VWR Item#MFLX06410-18
Your Organization's Price:
Retrieving
Resists chemicals and UV-light.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 3/4" ID x 1" OD
This tubing is ideal for chemical, heat, ozone, and ultraviolet (UV) light resistance applications. Characteristics include nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. They are manufactured according to GMP. Temperature range is -75 to 275 °F (-60 to 135 °C). Can be sterilized with autoclave.