Masterflex® Transfer Tubing, Versilon™ A-60-G, ¹⁄₁₆" ID × ³⁄₁₆" OD, 50 ft.
Supplier: VWR International
Resists chemicals and UV-light.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimize downtime
- Tubing size: 1/16" ID x 3/16" OD
This tubing is ideal for chemical, heat, ozone, and ultraviolet (UV) light resistance applications. Characteristics include nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. They are manufactured according to GMP. Temperature range is -75 to 275 °F (-60 to 135 °C). Can be sterilized with autoclave.