Masterflex® Transfer Tubing, Tygon® E-Lab (E-3603), 1/16" ID x 3/16" OD; 50 Ft
Supplier: VWR International
VWR Item#MFLX06407-72
Your Organization's Price:
Retrieving
Pliable formulation for sharp radius bending.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 1/16" ID x 3/16" OD
This tubing is ideal for general laboratory use. It offers long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, and is nontoxic. <br /><br />FDA compliant and made according to GMP. <br /><br />Temperature range is -51 to 165 °F (-46 to 73 °C)<br /><br />Can be sterilized with ethylene oxide or autoclave