Masterflex® Transfer Tubing, Tygon® E-Lab (E-3603), 1/32" ID x 3/32" OD; 50 Ft
Supplier: VWR International
Pliable formulation for sharp radius bending.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimize downtime
- Tubing size: 1/32" ID x 3/32" OD
This tubing is ideal for general laboratory use. It offers long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, and is nontoxic. <br /><br />FDA compliant and made according to GMP. <br /><br />Temperature range is -51 to 165 °F (-46 to 73 °C)<br /><br />Can be sterilized with ethylene oxide or autoclave