Fitting, PTFE, Straight, Threaded Plug, 1/4" NPT(M)
Supplier: MICROMOLD PRODUCTS, INC.
VWR Item#MFLX34030-56
Your Organization's Price:
Retrieving
Deigned for permanent connection.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/4" NPT(M)
Very good chemical resistance. Rated to 200 psi at 77 °F ( 25 °C). Usable from -65 to 190 °F (-53 to 87 °C).