Masterflex® L/S® 2-Stop Precision Pump Tubing, FDA-Compliant Viton®, L/S 13; 8/Pk
Supplier: VWR International
Good for food and lab applications.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique applicationUnique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Tubing Size: L/S 13
<B>Applications</B>: Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work<br /><B>Characteristics</B>: Good chemical resistance, opaque black<br />For use with Masterflex® multichannel pump heads.