Masterflex® Transfer Tubing, Black Rubber Vacuum, 3/4" ID x 1-1/2" OD; 25 ft
Supplier: VWR International
VWR Item#MFLX96143-17
Your Organization's Price:
Retrieving
Suitable for vacuum use.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: ³/₄" int.Ø × 1-¹/₂" out.Ø
The tubing is ideal for vacuum and pressure applications for general lab and industrial use. It features thick flexible walls with a temperature range of 0 to 150 °F (-18 to 65 °C). Can be sterilized with ethylene oxide or autoclave for 30 minutes at 15 psi (1.0 bar).