Masterflex® Transfer Tubing, Black Rubber Vacuum, 1/2" ID x 1" OD; 25 ft
Supplier: VWR International
Suitable for vacuum use.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimize downtime
- Tubing size: ¹/₂" int.Ø × 1" out.Ø
The tubing is ideal for vacuum and pressure applications for general lab and industrial use. It features thick flexible walls with a temperature range of 0 to 150 °F (-18 to 65 °C). Can be sterilized with ethylene oxide or autoclave for 30 minutes at 15 psi (1.0 bar).