Black Rubber Vacuum Tubing, 1/2" ID×1" OD; 25 ft
Supplier: VWR International
VWR Item#MFLX96143-10
Your Organization's Price:
Retrieving
Ideal for vacuum and pressure applications.
Applications: Vacuum and pressure applications for general lab and industrial use<br />Characteristics: Thick wall, flexible, opaque<br />Temperature Range: 0 °F to 150 °F<br />Sterilize: Ethylene oxide or autoclave for 30 minutes at 15 psi