Masterflex® Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 5/8" ID x 7/8" OD; 50 Ft
Supplier: VWR International
VWR Item#MFLX95633-09
Your Organization's Price:
Retrieving
No swelling or embrittlement.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 5/8" ID x 7/8" OD
The Tygon tubing is ideal for fuels and industrial lubricants—gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants. Resists most hydrocarbons, embrittlement and swelling, ozone- and UV-resistant, with low-extractability. Tubing is manufactured according to GMP and has a temperature range of -35 to 165 °F (-37 to 74 °C). Sterilization is not recommended.