Masterflex® Transfer Tubing, Polytetrafluoroethylene, 1/4" ID x 3/8" OD; 250 ft
Supplier: VWR International
VWR Item#MFLX95231-10
Your Organization's Price:
Retrieving
Available in spooled rolls for increased versatility.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: ¹/₄" int.Ø × ³/₈" out.Ø
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.