Masterflex® Ismatec® Footswitch for BVP and MCP Series Drives, DB-9 Male Connector; 6-ft (1.8 m)
Supplier: Avantor Fluid Handling
Simplify your dispensing.
- Select the footswitch with connectio suitable for your Ismatec® pump
Footswitches provide convenient, hands-free operation of your pump drives from up to six feet away. Simply press once to start pump, press again to stop.