Masterflex® Submersible Centrifugal Pump, 100 GPM/22 ft, 304 SS, High-Flow, Manual, 115 VAC
Supplier: VWR International
VWR Item#MFLX75505-57
Your Organization's Price:
Retrieving
Pump liquids with suspended solids up to 2" diameter at flow rates up to 175 GPM.
  • Stainless steel pumps are chemical and corrosion resistant
  • Manually-operated pumps allow for continuous emptying and automatically-operated pumps are used in on-demand dewatering applications
  • Dual-seal design ensures rugged and reliable service
  • Maximum flow rate 100 GPM
  • Maximum feet of head 22 ft
  • Operates on 115 VAC
Submersible pumps are ideal for food and beverage processing; irrigation and storm water runoff; spraying; and water and wastewater treatment applications. Operate pumps either fully or partially submerged.