Masterflex® High-Flow Dual Piston Pump, without Dampener, 10 mL/min, 316 SS; 100-240 VAC
Supplier: VWR International
VWR Item#MFLX74931-20
Your Organization's Price:
Retrieving
The right pump for your HPLC application.
  • Flow rates from 10 to 300 ml/min
  • Digital stepper motor ensures accuracy at low flow rates
  • Remotely control pump with a PC via RS-232 and micro USB
  • Self-flush feature washes piston with every stroke
  • Interactive keypad controls make programming at the pump
  • Maximum flow rate of 10 mL/minute
  • Analog remote controls standard on dual-head and isocratic pumps
<B>Single-Head Pumps</B> precisely meter and dispense chemicals, perfect for small-scale laboratory fluid transfers and HPLC applications. Small footprint saves bench and fume hood space.<br /><B>High-Flow Dual-Head Pumps</B> are ideally suited to Preparative
<b>Delivery information: </b>Includes detachable power supply. Dual-head and isocratic pump models also include a removable drip tray.