Masterflex® High-Flow Dual Piston Pump, 100 mL/min, PEEK fluid path; 100-240 VAC
Supplier: VWR International
The right pump for your HPLC application.
- Flow rates from 10 to 300 ml/min
- Digital stepper motor ensures accuracy at low flow rates
- Remotely control pump with a PC via RS-232 and micro USB
- Self-flush feature washes piston with every stroke
- Interactive keypad controls make programming at the pump
- Maximum flow rate of 100 mL/minute
- PEEK fluid path
- Includes pulse dampener
- Analog remote controls standard on dual-head and isocratic pumps
<B>Single-Head Pumps</B> precisely meter and dispense chemicals, perfect for small-scale laboratory fluid transfers and HPLC applications. Small footprint saves bench and fume hood space.<br /><B>High-Flow Dual-Head Pumps</B> are ideally suited to Preparative
<b>Delivery information: </b>Includes detachable power supply. Dual-head and isocratic pump models also include a removable drip tray.