Masterflex® Foot Switch, for Modular Touch-Screen Pumps
Supplier: VWR International
VWR Item#MFLX74906-92
Your Organization's Price:
Retrieving
Make the most of your Masterflex® metering pumps with these accessories.
  • Optimize performance of your Masterflex pumps
Choose from the following service kits and foot switches to meet your metering pump needs.
Used as a remote start/stop control device. Compatible with pumps 07582-00, 07582-10 and 07582-20.