Stenner Double Head Adjustable Mechanical Peristaltic Pump, 170 GPD, 115 VAC
Supplier: Avantor
Exceptional reliability and ease of service reduce maintenance and cost.
  • Polycarbonate corrosion-resistant housing
  • 24/7 continuous duty cycle
  • Mount pumps vertically or horizontally
<B>Single Head, Adjustable Mechanical Feed Rate Pumps</B> run reliably and consistently against 100 psi of pressure. These adjustable models feature a 20:1 turndown ratio with an adjustable rate in 2.5% increments from 5 to 100%.