Masterflex® Close-Coupled Centrifugal Pump, 20 GPM, 316 SS, Low-Flow, Self-Priming, TEVN; 115 VAC
Supplier: VWR International
Compact stainless steel pumps available for both DC and AC input power.
- 316 stainless steel is ideal for corrosion resistance when pumping many liquids
- 12 and 24 VDC versions available for many of the pumps
- 12-foot suction lift is possible with self-priming models
- Chemical-resistant design
- Maximum flow rate 20 GPM
- Maximum feet of head 24 ft
- Available with totally enclosed non-ventilated motor
The 316 SS pumps are ideal for many commercial and industrial applications where good chemical compatibility is needed for the bulk transfer of the system liquid. A carbon/ceramic/Viton® bellows mechanical seal is standard on all pumps. <br /><br />The stripped wires allow for easy integration within your system.
<b>Delivery information: </b>Includes stripped wire leads.