Masterflex® Close-Coupled Centrifugal Pump, 12 GPM, 316 SS, Low-Flow; 208-230 VAC
Supplier: VWR International
VWR Item#MFLX72021-14
Your Organization's Price:
Retrieving
Compact stainless steel pumps available for both DC and AC input power.
  • 316 stainless steel is ideal for corrosion resistance when pumping many liquids
  • 12 and 24 VDC versions available for many of the pumps
  • 12-foot suction lift is possible with self-priming models
  • Chemical-resistant design
  • Maximum flow rate 12 GPM
  • Maximum feet of head 14 ft
  • Available with totally enclosed non-ventilated motor
The 316 SS pumps are ideal for many commercial and industrial applications where good chemical compatibility is needed for the bulk transfer of the system liquid. A carbon/ceramic/Viton® bellows mechanical seal is standard on all pumps. <br /><br />The stripped wires allow for easy integration within your system.
<b>Delivery information: </b>Includes stripped wire leads.