Flowline Switch-Tek Vibration Level Switch, 12-36 VDC; 1/Each
Supplier: FLOWLINE
Don’t worry about buildup—automatically adjusts for optimal performance, alerts when cleaning is required.
- Constructed of Glass filled Ryton® and Viton®
SwitchTek vibration level switch is an excellent choice for more challenging application liquids, including those with light to medium coating or scaling-type characteristics. Liquid examples include light sodium hydroxide, brine and copper sulfate solutions.