Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Thread, 7/16" Hex x 1/8-27; 25/PK
Supplier: VWR International
VWR Item#MFLX45518-86
Your Organization's Price:
Retrieving
Easily transition between luer fittings and UNF or NPT threads.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Connection Type: Male Luer Lock to Threaded, ⁷/₁₆" He× × ¹/₈-27
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.