Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Thread Adapter, 10-32; 25/PK
Supplier: VWR International
VWR Item#MFLX45518-84
Your Organization's Price:
Retrieving
Easily transition between luer fittings and UNF or NPT threads.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Connection Type: Male Luer Lock to Threaded, 10-32
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.