Masterflex® Fitting, Polycarbonate, Straight, Male Luer Lock to Thread Adapter, 10-32; 25/Pk
Supplier: VWR International
Easily transition between luer fittings and UNF or NPT threads.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Connection Type: 10-32
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.