Hedland H605B-010 Water Flowmeter, 1.0-10 GPM, Brass, 1/2" NPT(F)
Supplier: BADGER METER INC
VWR Item#MFLX32509-75
Your Organization's Price:
Retrieving
Easy to read scale.
  • Direct reading
  • Install in any position
  • 360º Rotatable Guard/Scale
  • Easier ot Read Linear Scale
  • No Flow Straighteners or Special Piping Required
  • Insensitive to Shock and Vibration
  • Good Viscosity Stability
  • Temperature up to 240ºF
  • Accuracy ±2% Full Scale
  • Repeatability ±1%
  • Special Scales Available
Ideal for applications that are sensitive to shock and vibration.