GF Signet 3-2536-V0 Rotor-X Flow Sensor, PVDF/Hastelloy/PVDF for 0.5-4" Pipe
Supplier: BURT PROCESS EQUIPMENT
VWR Item#MFLX32500-10
Your Organization's Price:
Retrieving
Measure low flow rates down to 0.3 ft/second.
  • Eliminate cavitation and minimize pressure drop
Measure low flow rates in almost any size pipe. These sensors contain an open-core rotor with a small magnet in each of the four blades. As they rotate, the magnets pass a Hall effect sensor, which generates a linear frequency output that is proportional to the flow velocity. This signal travels up to 1000 feet without amplification.<br />The open-core design eliminates cavitation at flow velocities from 0.3 to 20 ft/second. Closed or solid paddle-wheel designs create air bubbles at these velocities, causing a nonlinear and nonrepeatable signal.<br /><br />Sensor materials resist most chemicals. All models have PVDF rotor and Viton® O-ring; and include a 25-ft connecting cable.<br />ROTOR-X low-flow sensors are compatible with all of our line-powered flow indicators, accumulators, and controllers. Mount in any type of pipe. Order installation fittings separately.