Masterflex® Variable-Area Flowmeter with High-Res Valve, Correlated Reading, 316 SS Housing and Fittings, 65-mm Scale, 3990 mL/min Air
Supplier: AALBORG INSTRUMENTS & CONTROL
VWR Item#MFLX32046-23
Your Organization's Price:
Retrieving