Masterflex® Fitting Clamp, PVDF, Sanitary Tri-Clamp®, 1" and 1-1/2"; 1/EA
Supplier: VWR International
Maintain sanitary conditions in your processes.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Sanitary Tri-Clamp holds together two sanitary fittings that are 1 and 1-1/2"
These sanitary clamps are compatible with Tri-Clamp® fittings. The PVDF material has excellent chemical resistance with low leachable characteristics.