Compression pipe adapter, Brass, 3/4", Each
Supplier: MIDWEST VALVE & FITTING INC
VWR Item#MFLX31412-80
Your Organization's Price:
Retrieving
Create a secure connection with compression fittings.
  • Constructed of durable brass
Good chemical resistance. Rated to 400 psi maximum at 77 °F ( 25 °C). Usable from -85 to 482 °F (-65 to 250 °C).