Compression pipe adapter, Brass, 5/16"×3/8", Each
Supplier: MIDWEST VALVE & FITTING INC
Create a secure connection with compression fittings.
- Constructed of durable brass
Good chemical resistance. Rated to 400 psi maximum at 77 °F ( 25 °C). Usable from -85 to 482 °F (-65 to 250 °C).