Masterflex® Fitting, Polyethylene, Straight, Male Threaded Adapter, 3/8" NPT(M); 100/PK
Supplier: VWR International
VWR Item#MFLX30627-28
Your Organization's Price:
Retrieving
Maintain a permanent connection.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/8" NPT(M)
  • Constructed of durable polyethylene
Threaded fittings provide a more permanent fluid path. Choose the size and material to meet your application needs.