Masterflex® Fittings, HDPE, Elbow, Hose Barb Union, 3/8" ID; 10/PK
Supplier: VWR International
VWR Item#MFLX30622-78
Your Organization's Price:
Retrieving
Quick and easy assembly without tools.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/8" ID
Create 90° bends in flexible tubing—without the kinks!