Masterflex® In-Line Strainer Bottom, Clear Nylon, Low-Profile
Supplier: SPECIALTY MFG CO
VWR Item#MFLX29595-63
Your Organization's Price:
Retrieving
Easily customize strainer system to suit your application.
  • Choose from either black polypropylene or clear nylon strainer bottoms
  • Use with other low-profile strainer components
Build your own economical filtration system to filter out particulates from your fluids. Available in three classes: low-profile, intermediate, and large. Modify or change components of the same class to meet the demands of your application.<br /><br />Low-Profile Strainers are ideal for small-scale low-volume filtration. Use in research applications, cleaning equipment, and laboratory equipment.<br />Intermediate Strainers feature a large filtering area in a compact size. Perfect for commercial and industrial pumps and agricultural and marine applications.<br />Large Strainers are primarily used in industrial and commercial industries. They feature increased flow rate and large filtration area.<br /><br />REQUIRED SYSTEM COMPONENTS—Order four components from the same class:<br />Strainer top<br />Strainer mesh<br />Strainer bottom<br />Gasket